Merck
  • Characterisation of UV-cured acrylate networks by means of hydrolysis followed by aqueous size-exclusion combined with reversed-phase chromatography.
Characterisation of UV-cured acrylate networks by means of hydrolysis followed by aqueous size-exclusion combined with reversed-phase chromatography.
Journal of chromatography. A (2006-11-23)
R Peters, V M Litvinov, P Steeman, A A Dias, Y Mengerink, R van Benthem, C G de Koster, Sj van der Wal, P Schoenmakers
ABSTRACT
UV-cured networks prepared from mixtures of di-functional (polyethylene-glycol di-acrylate) and mono-functional (2-ethylhexyl acrylate) acrylates were analysed after hydrolysis, by aqueous size-exclusion chromatography coupled to on-line reversed-phase liquid-chromatography. The mean network density and the fraction of dangling chain ends of these networks were varied by changing the concentration of mono-functional acrylate. The amount and the molar-mass distribution of the polyethylene-glycol chains between cross-links (M(XL)) and polyacrylic acid (PAA) backbone chains (the so-called kinetic chain length (kcl)) in the different acrylate networks were determined quantitatively. The molar-mass distribution of kcl revealed an almost linear dependence on the concentration of mono-functional acrylate. Analysis of the starting materials showed a significant concentration of mono-functional polyethylene-glycol acrylate. In combination with the analysis of the extractables of the UV-cured networks (polymers not attached to the network, impurities that originate from the photo-initiator and unreacted monomers), more insight in the total network structure was obtained. It was shown that the UV-cured networks contain only small fractions of residual compounds. With these results, the chemical network structure for the different UV-cured acrylate polymers was expressed in network parameters such as the number of PAA units which are cross-linked, the degree of cross-linking, and the network density, which is the molar concentration of effective network chains between cross-links per volume of the polymers. The mean molar mass of chains between chemical network junctions (M(C)) was calculated and compared with results obtained from solid-state NMR and DMA. The mean molar mass of chains between network junctions as determined by these methods was similar.
Sigma-Aldrich
2-Ethylhexyl acrylate, 98%, contains ≥0.001-≤0.11% monomethyl ether hydroquinone as stabilizer
Supelco
2-Ethylhexyl acrylate, analytical standard