Merck
  • [Effect of 2-methoxycinnamaldehyde on activity of COX and PGE2 release in cerebral microvascular endothelial cells stimulated by IL-1].
[Effect of 2-methoxycinnamaldehyde on activity of COX and PGE2 release in cerebral microvascular endothelial cells stimulated by IL-1].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2006-10-20)
Jian-you Guo, Yuan-xiao Yang, Bao-sheng Zhao, Hong-bin Liu, Lan-fang Li, Yue-Ying Ma, Shu-ying Guo, Hai-ru Huo, Ting-liang Jiang
ABSTRACT
To observe the effect of 2-methoxycinnamaldehyde (isolated from fraction A of Guizhi Tang) on activity of COX and PGE2 release in rat cerebral microvascular endothelial cells (rCMEC) stimulated by IL-1. rCMEC were cultured, and identified by immunohistochemistry for von Willebrand factor (VIII factor, a marker for all endothelial cells) in cytoplasm of the cells. Different concentrations of 2-methoxycinnamaldehyde were added respectively and incubated for 3 hours, then stimulated for another 12 hours by IL-1. Activities of COX-1 and COX-2 in rCMEC, and production of PGE2 in the conditioned media were measured by ELISA. Positive immunostaining for VIII factor was present diffusely in the cytoplasm of > 90% rCMEC. After being exposed to 30 ng x mL(-1) IL, the activity of COX-2 in rCMEC and the production of PGE2 in conditioned media were higher than those of control group, while there was no difference on activity of COX-1 in the two groups. 2-methoxycinnamaldehyde could down-regulate them in concentration-dependently, and significant differences on the activity of COX-2 and amount of PGE2 were showed in 200 microg x mL(-1) concentration. 2-methoxycinnamaldehyde can affect the PGE2 release in rCMEC induced by IL-1, which might be related with its inhibition on the activity of COX-2.
Sigma-Aldrich
2-Methoxycinnamaldehyde, natural, 98%, FG
Pricing and availability is not currently available.