Merck
Ozonation of acid yellow 17 dye in a semi-batch bubble column.
Journal of hazardous materials (2006-08-15)
Laura W Lackey, Richard O Mines, Philip T McCreanor
ABSTRACT
A semi-batch bubble column was used to evaluate the effect of ozonation on the removal of acid yellow 17 dye from water. Results indicate that ozonation is very effective at removing acid yellow 17 dye from synthetic textile wastewater. The ozone consumed to apparent dye removal ratio ranged from 2 to 15,000 mg ozone per mg of dye decolorized and was dependent on both ozonation time and apparent dye concentration. The biodegradability of the dye wastewater was evaluated by monitoring changes in 5-day biochemical oxygen demand (BOD5) with respect to chemical oxygen demand (COD). Results indicate that the wastewater biodegradability increased with an increase in ozonation time. Film theory was used to kinetically model the gas-liquid reactions occurring in the reactor. Modeling results indicated that during the first 10-15 min of ozonation, the system could be characterized by a fast, pseudo-first-order regime. With continued ozonation, system kinetics transitioned through a moderate then to a slow regime. Successful modeling of this period required use of a kinetic equation corresponding to a more inclusive condition. Model results are presented.
Product Description
Supelco
Yellow 2G, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Acid Yellow 17, Dye content 60 %
Pricing and availability is not currently available.