Merck
  • [Calcium carbide of different crystal formation synthesized by calcium carbide residue].
[Calcium carbide of different crystal formation synthesized by calcium carbide residue].
Huan jing ke xue= Huanjing kexue (2006-06-14)
Zhong-yuan Lu, Ming Kang, Cai-rong Jiang, Ming-jing Tu
ABSTRACT
To recycle calcium carbide residue effectively, calcium carbide of different crystal form, including global aragonite, calcite and acicular calcium carbide was synthesized. Both the influence of pretreatment in the purity of calcium carbide, and the influence of temperatures of carbonization reaction, release velocity of carbon dioxide in the apparition of calcium carbide of different crystal form were studied with DTA-TG and SEM. The result shows that calcium carbide residue can take place chemistry reaction with ammonia chlorinate straight. Under the condition that pH was above 7, the purity of calcium carbide was above 97%, and the whiteness was above 98. Once provided the different temperatures of carbonization reaction and the proper release velocity of carbon dioxide, global aragonite, calcite and acicular calcium carbide were obtained.
Sigma-Aldrich
Calcium carbide, CP
Sigma-Aldrich
Calcium carbide, granulated, technical, ≥75% (gas-volumetric)
Sigma-Aldrich
Calcium carbide, pieces, thickness <10 mm , typically, technical grade, ~80%