- Synthesis, isolation and characterization of cationic gold(I) N-heterocyclic carbene (NHC) complexes.
Synthesis, isolation and characterization of cationic gold(I) N-heterocyclic carbene (NHC) complexes.
A number of cationic gold(I) complexes have been synthesized and found to be stabilized by the use of N-heterocyclic carbene ligands. These species are often employed as in situ-generated reactive intermediates in gold catalyzed organic transformations. An isolated, well-defined species was tested in gold-mediated carbene transfer reactions from ethyl diazoacetate.