Merck
Functionalisation of carbon nanohorns.
Chemical communications (Cambridge, England) (2006-05-17)
Carla Cioffi, Stéphane Campidelli, Fulvio G Brunetti, Moreno Meneghetti, Maurizio Prato
ABSTRACT
The functionalisation of single wall carbon nanohorns via 1,3-dipolar cycloaddition has been achieved, and the products have been characterised by spectroscopy, microscopy and thermogravimetry.
Sigma-Aldrich
Heptaldehyde, 95%
Pricing and availability is not currently available.
Sigma-Aldrich
Heptanal, ≥95%, FCC, FG
Pricing and availability is not currently available.