Merck
Release of iron from ferritin requires lysosomal activity.
American journal of physiology. Cell physiology (2006-04-14)
Theodros Z Kidane, Eric Sauble, Maria C Linder
ABSTRACT
How ferritin-Fe becomes available for cell functions is unknown. Our previous studies with rat hepatoma cells indicated ferritin had to be degraded to release its Fe. In these studies, we investigated whether this occurs in other cell types and whether lysosomes are required. Release of ferritin-Fe was induced with desferoxamine (DFO) in (59)Fe-preloaded hepatoma, Caco2, and erythroid K562 cells and measured by rocket immunoelectrophoresis and autoradiography. The half-lives for ferritin-(59)Fe and protein were parallel (23, 16, and 11 h for the hepatic, Caco2, and K562 cells, respectively). Co-treatment with 180 microM Fe, leupeptin, chymostatin, or chloroquine markedly decreased rates of ferritin-Fe release and ferritin degradation. Lactacystin had no effect except for a small one in erythroid cells. Fractionation of hepatoma cell lysates on iodixanol gradients showed rapid depletion of cytosolic ferritin by DFO treatment but no accumulation in lysosomes. We conclude that regardless of cell type, release of Fe from ferritin occurs mainly through lysosomal proteolysis.
Sigma-Aldrich
Chymostatin, microbial
Pricing and availability is not currently available.