- Determination of ceftriaxone sodium in pharmaceutical formulations by flow injection analysis with acid potassium permanganate chemiluminescence detection.
Determination of ceftriaxone sodium in pharmaceutical formulations by flow injection analysis with acid potassium permanganate chemiluminescence detection.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2006-01-25)
Deyi Zhang, Yongjun Ma, Min Zhou, Li Li, Hui Chen
PMID16429800
ABSTRACT
Based on the chemiluminescence (CL) emission generated from the oxidation of ceftriaxone sodium alkali hydrolysate by potassium permanganate in polyphosphoric acid (PPA), a novel determination method for ceftriaxone sodium was developed by using a flow-injection technique. The calibration curve appears to be linear in the range between 0.05 and 100 microg mL(-1) with a detection limit (3sigma) of 25 ng mL(-1), and a relative standard deviation (RSD) of 0.6% for eleven replicate determinations of 5.0 microg mL(-1) ceftriaxone sodium. The proposed method has been successfully utilized for the determination of ceftriaxone sodium in pharmaceutical formulations, while the chemiluminescence reaction mechanisms were investigated.