Merck
  • 2-Ene-1,4-diols by dimerization of terminal epoxides using hindered lithium amides.
2-Ene-1,4-diols by dimerization of terminal epoxides using hindered lithium amides.
Organic letters (2005-06-04)
David M Hodgson, Christopher D Bray, Nicholas D Kindon
ABSTRACT
[reaction: see text] Reaction of hindered lithium amides with readily available (enantiopure) terminal epoxides gives 2-ene-1,4-diols via carbenoid dimerization of the corresponding alpha-lithiated epoxides. D-Mannitol and D-iditol were synthesized using this method in three steps from (S)-tritylglycidyl ether.
Sigma-Aldrich
L-Iditol, ≥98% (GC)
Pricing and availability is not currently available.