Merck
  • Solvent-free condensation of phenylacetonitrile and nonanenitrile with 4-methoxybenzaldehyde: optimization and mechanistic studies.
Solvent-free condensation of phenylacetonitrile and nonanenitrile with 4-methoxybenzaldehyde: optimization and mechanistic studies.
Organic & biomolecular chemistry (2005-04-14)
André Loupy, Michèle Pellet, Alain Petit, Giang Vo-Thanh
ABSTRACT
The condensation of phenylacetonitrile with 4-methoxybenzaldehyde can be carried out by two solvent-free methods: i) using neat powdered KOH at room temperature with equivalent amounts of aldehyde, nitrile and base for 3 min, and ii) using K2CO3 in the presence of a phase transfer agent for 3 min under microwave activation or conventional heating. By extending the reaction time up to 10 min, four different products were obtained from phenyl or nitrile group migration. With nonanenitrile, only the second method could be applied to give two kinds of condensed compounds with or without phenyl (or nitrile) group migration. The intervention of non-thermal MW-specific effects was proved in some cases and interpreted.
Sigma-Aldrich
Benzyl cyanide, 98%
Sigma-Aldrich
p-Anisaldehyde, 98%
Supelco
p-Anisaldehyde, analytical standard