Merck
  • Use of biotinylated 17beta-estradiol in enzyme-immunoassay development: spacer length and chemical structure of the bridge are the main determinants in simultaneous streptavidin-antibody binding.
Use of biotinylated 17beta-estradiol in enzyme-immunoassay development: spacer length and chemical structure of the bridge are the main determinants in simultaneous streptavidin-antibody binding.
Journal of immunological methods (2005-03-22)
Markus Lacorn, Kerstin Fleischer, Stephanie Willig, Sabine Gremmel, Hans Steinhart, Rolf Claus
ABSTRACT
17beta-estradiol (E2) concentrations are in the low pg/ml range in plasma. To develop a sensitive enzyme immunoassay (EIA) for E2-determination a highly specific antibody raised against a 6-carboxymethyl (CMO)-E2-bovine serum albumine conjugate was used. Based on 6-CMO-E2 and 6-amino-E2, four biotinylated tracers with two different spacer lengths between E2 and biotin were synthesized using biotinylation reagents in one step reactions. All amino-based tracers were unsuitable for assay development because the antibody binding was too weak compared to the analyte E2. For 6-CMO-based tracers the simultaneous binding of the tracer to the antibody and streptavidin seems to be the determining step in the procedure depending on incubation temperature and spacer lengths. While a short spacer of 9 carbon atoms was susceptible to room temperature, a longer spacer of 16 carbon atoms showed nearly the same results for incubation at 4 degrees C or at room temperature. The absolute detection limit of this system was 0.63 pg/well. For sample clean-up, porcine plasma was solvent-extracted and depending on the initial plasma volume further purified by solvent partition. Determination of reproducibility resulted in intraassay coefficients of variation of 13% and 5.3% for samples with E2-levels of 15 pg/ml and 236 pg/ml, respectively. Measurement of E2-spiked blood plasma revealed recoveries of 83% up to 100% for E2 concentrations between 50 pg/ml and 1000 pg/ml. Only for the lowest concentration (20 pg/ml) a recovery of 58% was observed. Correlation of the EIA with an established radio immunoassay resulted in r=0.991 using the same antibody.
Sigma-Aldrich
β-Estradiol-6-one 6-(O-carboxymethyloxime)