Merck
  • A new isolate of Pseudomonas stutzerithat degrades 2-chloronitrobenzene.
A new isolate of Pseudomonas stutzerithat degrades 2-chloronitrobenzene.
Biotechnology letters (2005-03-03)
Hong Liu, Shu-Jun Wang, Ning-Yi Zhou
ABSTRACT
A strain of Pseudomonas stutzeri ZWLR2-1 was isolated from soil contaminated with chloronitrobenzenes and identified by 16S rDNA sequencing. This bacterium released chloride and nitrite into the medium when grown on 0.5 mM 2-chloronitrobenzene. PCR amplification and DNA sequencing revealed a DNA fragment encoding a polypeptide homologous to the alpha-subunit of ring-hydroxylating dioxygenases.
Sigma-Aldrich
1-Chloro-2-nitrobenzene, 99%