Merck
Enantioselective route from carbohydrates to cyclooctane polyols.
Organic letters (2005-01-28)
Leo A Paquette, Yunlong Zhang
ABSTRACT
[reaction: see text] A synthetic route to select cyclooctane-1,2,3-triols and 1,2,3,4,5-pentaols has been defined. The starting materials are d-glucose or d-arabinose, and the key steps consist of a zirconocene-promoted ring contraction, a [3,3] sigmatropic rearrangement, and more extended functionalization of the resulting cyclooctadienone.
Sigma-Aldrich
Bis(cyclopentadienyl)zirconium(IV) dichloride, ≥98%