- Aphrodisiac drug-induced hemolysis.
Aphrodisiac drug-induced hemolysis.
Journal of toxicology. Clinical toxicology (2004-09-15)
Ruth Stalnikowicz, Yona Amitai, Yedidia Bentur
PMID15362601
ABSTRACT
Volatile alkyl nitrites have been used during the past decades for "recreational purposes," and for intensifying sexual experience. Their use has been associated with methemoglobinemia and hemolysis. We report three patients who presented over the past year with acute hemolysis after inhalation of butyl nitrite, two of them had glucose-6-phosphate dehydrogenase (G6PD) deficiency.