Merck
  • Interrogating conformationally dependent electron-transfer dynamics via ultrafast visible pump/IR probe spectroscopy.
Interrogating conformationally dependent electron-transfer dynamics via ultrafast visible pump/IR probe spectroscopy.
Journal of the American Chemical Society (2004-03-05)
Igor V Rubtsov, Naomi P Redmore, Robin M Hochstrasser, Michael J Therien
ABSTRACT
We demonstrate for the first time the utility of time-resolved visible pump/mid-infrared (IR) probe spectroscopy to interrogate directly, and provide unique infomation regarding, conformationally dependent photoinduced ET dynamics and the subsequent structural evolution of the resulting charge-separated (CS) state. Exemplary polarized visible pump/IR probe experiments involving N-[5-(10,20-diphenylporphinato)zinc(II)]-N'(octyl)pyromellitic diimide (PZn-PI) and [5-[4'-(N-(N'-octyl)pyromellitic diimide)phenyl)ethynyl]-10,20-diphenylporphinato]zinc(II) (PZn()PI) show that it is possible to assess the mean PZn-to-PI interplanar torsional angle of electronically excited structural conformers that undergo ET within the sub-ps time domain for both of these donor-spacer-acceptor (D-Sp-A) systems. Further transient specroscopic experiments carried out on PZn()PI determine how this angle evolves with time. Because vibrational transition moments are often known and typically localized, this work underscores that polarized visible pump/IR probe spectroscopy defines a valuable tool to interrogate structrure in both electronically excited and CS states; this fact, coupled with the ultrafast time resolution and high senseitivity offered by this technique, make it ideally suited to probe a range of mechanistic issues relevant to charge-transfer reactions.
Sigma-Aldrich
Pyromellitic diimide, 97%
Pricing and availability is not currently available.