Merck
  • Synthesis of novel carboacyclic nucleosides with vinyl bromide moiety as open-chain analogues of neplanocin A.
Synthesis of novel carboacyclic nucleosides with vinyl bromide moiety as open-chain analogues of neplanocin A.
Archives of pharmacal research (2004-01-16)
Myung-Hee Choi, Hee-Doo Kim
ABSTRACT
A novel carboacyclic nucleoside analogue, 9-[2-bromo-4-hydroxy-3-hydroxymethyl-2-butenyl] adenine, and its derivatives were designed and synthesized as open-chain analogues of neplanocin A. The syntheses were accomplished via the coupling of adenine or pyrimidine bases to the key intermediate allylic bromide 7. The bromide 7 was prepared from epichlorohydrin in a seven step process in a 54% overall yield. The synthesized compounds were evaluated for their antiviral activity against the polio virus, HSV and HIV.
Sigma-Aldrich
Vinyl bromide solution, 1.0 M in THF
Sigma-Aldrich
Vinyl bromide, 98%