Merck
  • Effect of dietary selenite on N-nitrosodiethylamine-induced and phenobarbital promoted multistage hepatocarcinogenesis in rat: reflection in some minerals.
Effect of dietary selenite on N-nitrosodiethylamine-induced and phenobarbital promoted multistage hepatocarcinogenesis in rat: reflection in some minerals.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2003-12-04)
C Thirunavukkarasu, D Sakthisekaran
ABSTRACT
Selenium (Se), a dietary micronutrient, plays a vital role in cancer chemotherapy in many organs including the liver. We have studied the relationship between some minerals, which are essential in normal functioning of cells and anticancer effect of Se in N-nitrosodiethylamine (DEN) induced and phenobarbital (PB) promoted multistage hepatocarcinogenesis. Se (4 ppm through drinking water; as sodium selenite) was given to animals throughout the study, before initiation and during promotion phase of hepatocarcinogenesis, in a defined experimental protocol. Se, sodium, potassium, calcium and iron were measured either in hepatoma, or surrounding liver tissue or whole liver tissue and serum of experimental animals. DEN and PB treatment significantly (P < 0.001) increased potassium, calcium and iron levels in serum, while it decreased (P < 0.001) the Se and sodium levels when compared with control rats. We have also observed significantly increased (P < 0.001) sodium, calcium and iron levels in hepatoma and surrounding liver tissue, whereas, Se, and potassium level was found to be decreased (P < 0.001) when compared with control rats. Supplementation of selenite throughout the study, before initiation and during promotion stage significantly alters the above mineral content. Results showed that the most significant beneficial effect of selenium during hepatocarcinogenesis was exerted potentially in long-term continuous and/or before the initiation phase of carcinogenicity, rather than in the promotion phase. The present and previous results from our laboratory suggest that sub-optimal intake of a single trace mineral can have broad effects on chemotherapy, providing a framework for understanding the multiple beneficial effects of selenium in cancer chemoprevention.