Merck
  • Mixed-mode electrokinetic chromatography of aromatic bases with two pseudo-stationary phases and pH control.
Mixed-mode electrokinetic chromatography of aromatic bases with two pseudo-stationary phases and pH control.
Journal of chromatography. A (2003-07-02)
Philip Zakaria, Miroslav Macka, Paul R Haddad
ABSTRACT
The electrokinetic chromatographic (EKC) separation of a series of aromatic bases was achieved utilising an electrolyte system comprising an anionic soluble polymer (polyvinylsulfonic acid, PVS) and a neutral beta-cyclodextrin (beta-CD) as pseudo-stationary phases. The separation mechanism was based on a combination of electrophoresis, ion-exchange interactions with PVS, and hydrophobic interactions with beta-CD. The extent of each chromatographic interaction was independently variable, allowing for control of the separation selectivity of the system. The ion-exchange and the hydrophobic interactions could be varied by changing the PVS and the beta-CD concentrations, respectively. Additionally, mobilities of the bases could be controlled by varying pH, due to their large range of pKa values. The separation system was very robust with reproducibility of migration times being <2% RSD. The two-dimensional parameter space defined by the two variables, [beta-CD] and %PVS, was modelled using a physical model derived from first principles. This model gave very good correlation between predicted and observed mobilities (r2=0.999) for the 13 aromatic bases and parameters derived from the model agreed with the expected ion-exchange and hydrophobic character of each analyte. The complexity of the mathematical model was increased to include pH and this three-dimensional system was modelled successfully using an artificial neural network (ANN). Optimisation of both the two-dimensional and three-dimensional systems was achieved using the normalised resolution product and minimum resolution criteria. An example of using the ANN to predict conditions needed to obtain a separation with a desired migration order between two of the analytes is also shown.
Sigma-Aldrich
Poly(vinylsulfonic acid, sodium salt) solution, 30-40 wt. % in H2O, technical grade
Pricing and availability is not currently available.