Merck
  • Studies on the role of some synthetic curcuminoid derivatives in the inhibition of tumour specific angiogenesis.
Studies on the role of some synthetic curcuminoid derivatives in the inhibition of tumour specific angiogenesis.
Journal of experimental & clinical cancer research : CR (2003-05-03)
P V Leyon, G Kuttan
ABSTRACT
In this study, some of the synthetic curcuminoid derivatives are analyzed for their anti-angiogenic activity. Intraperitoneal administration of the compounds tetrahydrocurcumin (THC), salicyl curcumin (SC) and curcuminIII (C-III) reduced the number of tumour directed capillaries induced by injecting B16F-10 melanoma cells on the ventral side of C57BL/6 mice. THC (14.5 +/- 2.5 capillaries) and SC (16 +/- 2.5 capillaries) were more significant (P < 0.001) than C-III (19 +/- 1.8 capillaries) compared to the untreated control (30.8 +/- 2.1 capillaries). Treatment with these curcuminoids reduced the serum NO as well as TNF-alpha levels of the angiogenesis-induced animals. In vitro NO and TNF-alpha production by the activated MOs were reduced in a concentration dependent manner by the treatment of the curcuminoid compounds.
Product Description
Supelco
Bisdemethoxycurcumin, analytical standard
Pricing and availability is not currently available.