Merck
  • 4-nitrocatechol as a probe of a Mn(II)-dependent extradiol-cleaving catechol dioxygenase (MndD): comparison with relevant Fe(II) and Mn(II) model complexes.
4-nitrocatechol as a probe of a Mn(II)-dependent extradiol-cleaving catechol dioxygenase (MndD): comparison with relevant Fe(II) and Mn(II) model complexes.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2003-02-18)
Mark F Reynolds, Miquel Costas, Masami Ito, Du-Hwan Jo, A Alex Tipton, Adam K Whiting, Lawrence Que
ABSTRACT
Mn(II)-dependent 3,4-dihydroxyphenylacetate 2,3-dioxygenase (MndD) is an extradiol-cleaving catechol dioxygenase from Arthrobacter globiformis that has 82% sequence identity to and cleaves the same substrate (3,4-dihydroxyphenylacetic acid) as Fe(II)-dependent 3,4-dihydroxyphenylacetate 2,3-dioxygenase (HPCD) from Brevibacterium fuscum. We have observed that MndD binds the chromophoric 4-nitrocatechol (4-NCH(2)) substrate as a dianion and cleaves it extremely slowly, in contrast to the Fe(II)-dependent enzymes which bind 4-NCH(2) mostly as a monoanion and cleave 4-NCH(2) 4-5 orders of magnitude faster. These results suggest that the monoanionic binding state of 4-NC is essential for extradiol cleavage. In order to address the differences in 4-NCH(2) binding to these enzymes, we synthesized and characterized the first mononuclear monoanionic and dianionic Mn(II)-(4-NC) model complexes as well as their Fe(II)-(4-NC) analogs. The structures of [(6-Me(2)-bpmcn)Fe(II)(4-NCH)](+), [(6-Me(3)-TPA)Mn(II)(DBCH)](+), and [(6-Me(2)-bpmcn)Mn(II)(4-NCH)](+) reveal that the monoanionic catecholate is bound in an asymmetric fashion (Delta r(metal-O(catecholate))=0.25-0.35 A), as found in the crystal structures of the E(.)S complexes of extradiol-cleaving catechol dioxygenases. Acid-base titrations of [(L)M(II)(4-NCH)](+) complexes in aprotic solvents show that the p K(a) of the second catecholate proton of 4-NCH bound to the metal center is half a p K(a) unit higher for the Mn(II) complexes than for the Fe(II) complexes. These results are in line with the Lewis acidities of the two divalent metal ions but are the opposite of the trend observed for 4-NCH(2) binding to the Mn(II)- and Fe(II)-catechol dioxygenases. These results suggest that the MndD active site decreases the second p K(a) of the bound 4-NCH(2) relative to the HPCD active site.
Sigma-Aldrich
4-Nitrocatechol, ≥96.0%