Merck
  • Induction of birth in the bandicoot (Isoodon macrourus) with prostaglandin and oxytocin.
Induction of birth in the bandicoot (Isoodon macrourus) with prostaglandin and oxytocin.
Reproduction (Cambridge, England) (2002-02-28)
R T Gemmell
ABSTRACT
As in eutherians, maturation of the fetal pituitary and adrenal glands together with an increase in prostaglandin and mesotocin or oxytocin production initiates birth in marsupials. In this study, prostaglandin (Lutalyse) or oxytocin (Syntocinon) were administered to pregnant bandicoots at 05:00 h on the calculated day of birth and the resultant effects were filmed for analysis. The administration of prostaglandin caused the bandicoot to adopt the birth position several minutes after injection (n = 2). However, the bandicoot did not give birth for several hours. Birth occurred at a similar time of day to that observed for untreated bandicoots (n = 7), between 08:00 h and 12:00 h. After an injection of oxytocin, the bandicoot assumed the birth position and birth occurred within several minutes. The young were alive while still connected to their allantoic stalks. However, they were unable to attach to the teats and did not survive (n = 4). The induced young were the colour of venous blood and died soon after the umbilicus was separated, indicating that the cardiopulmonary system of these neonates was underdeveloped and inadequate to maintain life. The results from this study demonstrate that prostaglandin is required to prepare the bandicoot for birth, and mesotocin is required for contraction of the uterus and for birth to occur.
Sigma-Aldrich
Prostaglandin F tris salt, ≥99%, synthetic, powder
Sigma-Aldrich
Prostaglandin F tris salt, synthetic, suitable for cell culture