Merck
  • Structure and cooperativity of a T-state mutant of histidine decarboxylase from Lactobacillus 30a.
Structure and cooperativity of a T-state mutant of histidine decarboxylase from Lactobacillus 30a.
Proteins (2002-02-09)
Scott Worley, Elisabeth Schelp, Arthur F Monzingo, Stephen Ernst, Jon D Robertus
ABSTRACT
Histidine decarboxylase (HDC) from Lactobacillus 30a converts histidine to histamine, a process that enables the bacteria to maintain the optimum pH range for cell growth. HDC is regulated by pH; it is active at low pH and inactive at neutral to alkaline pH. The X-ray structure of HDC at pH 8 revealed that a helix was disordered, resulting in the disruption of the substrate-binding site. The HDC trimer has also been shown to exhibit cooperative kinetics at neutral pH, that is, histidine can trigger a T-state to R-state transition. The D53,54N mutant of HDC has an elevated Km, even at low pH, indicating that the enzyme assumes the low activity T-state. We have solved the structures of the D53,54N mutant at low pH, with and without the substrate analog histidine methyl ester (HME) bound. Structural analysis shows that the apo-D53,54N mutant is in the inactive or T-state and that binding of the substrate analog induces the enzyme to adopt the active or R-state. A mechanism for the cooperative transition is proposed.
Sigma-Aldrich
L-Histidine methyl ester dihydrochloride, 97%
Pricing and availability is not currently available.