Merck
  • NOESY on neurotoxins: NMR and conformational assignments of picrotoxins.
NOESY on neurotoxins: NMR and conformational assignments of picrotoxins.
Phytochemical analysis : PCA (2001-11-14)
N B Perry, M Aiyaz, D S Kerr, R J Lake, M T Leach
ABSTRACT
Nuclear Overhauser effect spectroscopy (NOESY) gave full assignments of the 1H-NMR spectra of the picrotoxane neurotoxins tutin, hyenanchin, picrotoxinin and picrotin, as well as the solution conformations of these compounds, consistent with molecular modelling. Fully assigned 13C-NMR data are reported.
Sigma-Aldrich
Picrotoxinin
Pricing and availability is not currently available.