Merck
A fluoride-releasing composite for dental applications.
Dental materials : official publication of the Academy of Dental Materials (2001-02-13)
E A Glasspoole, R L Erickson, C L Davidson
ABSTRACT
The aim of this work was to attempt formulation of a dental composite, for investigational purposes, which will release fluoride at a rate comparable to glass ionomer materials, while retaining physical properties adequate for potential use in a number of dental applications. An organic fluoride material, tetrabutylammonium tetrafluoroborate (TBATFB), was incorporated into a hydrophilic monomer system made of 2,2-bis[4-(2-hydroxy-3-methacroyloxypropoxy)phenyl]-propane (BisGMA) and 2-hydroxyethyl methacrylate (HEMA). This fluoride-containing monomer system was filled with silane treated quartz to 81 wt%. The resultant material was tested for fluoride release, physical strength properties and bond strength to enamel, each in comparison to control materials. Cumulative fluoride release from the experimental composite was approximately linear over time and comparable to glass ionomer materials. The long term rate of release exceeded several of the glass ionomer materials tested. Diametral tensile strength was comparable to resin-modified glass ionomer (RMGI) materials. Bond strength to etched enamel was statistically equivalent to a commercially available hybrid composite control. The experimental resin-based composite material could potentially be used as an alternative to glass ionomer materials in non-stress bearing restorative applications and for orthodontic bracket bonding, where high fluoride release is desirable.
Sigma-Aldrich
Tetrabutylammonium tetrafluoroborate, 99%
Pricing and availability is not currently available.
Supelco
Tetrabutylammonium tetrafluoroborate, for electrochemical analysis, ≥99.0%
Pricing and availability is not currently available.