Merck
  • Fullerene lipids: synthesis of C60 fullerene derivatives bearing a long-chain saturated or unsaturated triester system.
Fullerene lipids: synthesis of C60 fullerene derivatives bearing a long-chain saturated or unsaturated triester system.
Lipids (1999-12-22)
M S Jie, S W Cheung
ABSTRACT
Tris(hydroxymethyl)aminomethane was successfully esterified with saturated and unsaturated long-chain fatty acids. The resulting amino-triester intermediates were successively reacted with chloroacetyl chloride, sodium azide, and C60 fullerene. Spectral evidence showed that the aziridine ring is joined to the junction of 16,6]-fused rings of the fullerene. The structures of the various C60 fullerene derivatives bearing a long-chain saturated or unsaturated triester system were characterized by spectroscopic and spectrometric methods.
Sigma-Aldrich
Chloroacetyl chloride, 98%