Merck
  • Determination of thiodiglycolic acid in urine by capillary electrophoresis.
Determination of thiodiglycolic acid in urine by capillary electrophoresis.
Journal of chromatography. A (1999-08-04)
E Samcová, V Kvasnicová, J Urban, I Jelínek, P Coufal
ABSTRACT
A sensitive capillary electrophoretic method was developed for the determination of thiodiglycolic acid (TDA) in urine which avoids the pretreatment of the urine sample. Several carrier electrolytes were examined. The most suitable carrier electrolyte system consisted of potassium hydrogen phthalate (5 mM), 2-(N-morpholino)ethanesulfonic acid (50 mM) and tetradecyltrimethylammonium bromide (0.5 mM), pH 5.2. Ten times diluted fresh midstream void urine was used for the determination. In this way, the concentrations of TDA between 5 and 50 mg/l in undiluted urine samples can be determined.
Sigma-Aldrich
2,2′-Thiodiacetic acid, 98%
Pricing and availability is not currently available.