Merck
Analysis of cell lineage relationships in taste buds.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2002-06-01)
Leslie M Stone, Seong-Seng Tan, Patrick P L Tam, Thomas E Finger
ABSTRACT
Taste buds are a heterogeneous population of cells exhibiting diverse morphological and biochemical characteristics. Because taste buds arise from multiple progenitors, the different types of taste cells may represent distinct lineages. The present study was undertaken to determine the following: (1) how many progenitors contribute to a taste bud, and (2) whether the specific subpopulation of serotonin-immunoreactive (IR) taste cells are related by lineage to a restricted set of progenitor cells. These questions were addressed using cell lineage analysis of taste buds from H253 X-inactivation mosaic mice. After random X-inactivation of the lacZ transgene, the tongue of hemizygous female mice displays discrete patches of epithelial cells, which are either beta-galactosidase (beta-gal) positive or beta-gal negative. By analyzing the proportion of the two differently stained cell populations in taste buds located at the boundary between positive and negative epithelial patches, we can determine the minimum number of progenitors that may contribute to the formation of a taste bud. The presence of taste buds containing only 6-12% labeled cells indicates that at least eight progenitors contribute to an average taste bud of 55 cells, assuming progenitors contribute equally to the cell population. Cell lineage analysis of serotonin-IR taste cells in such mixed taste buds suggests that this subpopulation likely arises from only one to two progenitors and often is related by lineage. Thus, at least some of the cell types in a taste bud represent distinct lineages of cells and are not merely phenotypic stages as a cell progresses from a young to a mature state.
Sigma-Aldrich
Bluo-Gal, reagent for selection of recombinant bacterial clones
Pricing and availability is not currently available.