Merck
  • IgG4 production is confined to human IL-10-producing regulatory B cells that suppress antigen-specific immune responses.
IgG4 production is confined to human IL-10-producing regulatory B cells that suppress antigen-specific immune responses.
The Journal of allergy and clinical immunology (2013-03-05)
Willem van de Veen, Barbara Stanic, Görkem Yaman, Marcin Wawrzyniak, Stefan Söllner, Deniz G Akdis, Beate Rückert, Cezmi A Akdis, Mübeccel Akdis
ABSTRACT
IL-10-producing regulatory B cells suppress immune responses, and lack of these cells leads to exacerbated symptoms in mouse models of chronic inflammation, transplantation, and chronic infection. IgG4 is a blocking antibody isotype with anti-inflammatory potential that is induced in human high-dose antigen tolerance models. We sought to characterize human inducible IL-10-secreting B regulatory 1 (BR1) cells and to investigate their immunoregulatory capacity through suppression of cellular immune responses and production of anti-inflammatory immunoglobulins. Highly purified IL-10-secreting B cells were phenotypically and functionally characterized by means of whole-genome expression analysis, flow cytometry, suppression assay, and antibody production. B cells specific for the major bee venom allergen phospholipase A2 (PLA) were isolated from beekeepers who displayed tolerance to bee venom antigens and allergic patients before and after specific immunotherapy. Human IL-10+ BR1 cells expressed high surface CD25 and CD71 and low CD73 levels. Sorting of CD73-CD25+CD71+ B cells allowed enrichment of human BR1 cells, which produced high levels of IL-10 and potently suppressed antigen-specific CD4+ T-cell proliferation. IgG4 was selectively confined to human BR1 cells. B cells specific for the major bee venom allergen PLA isolated from nonallergic beekeepers show increased expression of IL-10 and IgG4. Furthermore, the frequency of IL-10+ PLA-specific B cells increased in allergic patients receiving allergen-specific immunotherapy. Our data show the characterization of IL-10+ BR1 cells and in vivo evidence for 2 essential features of allergen tolerance: the suppressive B cells and IgG4-expressing B cells that are confined to IL-10+ BR1 cells in human subjects.
Sigma-Aldrich
Phospholipase A2 from honey bee venom (Apis mellifera), salt-free, lyophilized powder, 600-2400 units/mg protein
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phospholipase A2 from porcine pancreas, ammonium sulfate suspension, ≥600 units/mg protein
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phospholipase A2 from bovine pancreas, lyophilized powder, ≥20 units/mg protein