- Protease-mediated ligation of abiotic oligomers.
Protease-mediated ligation of abiotic oligomers.
Journal of the American Chemical Society (2005-12-08)
Barney Yoo, Kent Kirshenbaum
PMID16332030
ABSTRACT
We demonstrate that proteases can catalyze the ligation of peptidomimetic oligomers. The enzyme clostripain was used to facilitate the native ligation of N-substituted glycine oligomers, or peptoids. In addition to mediating the efficient condensation of two peptoid fragments, iterative ligation events were also performed, giving rise to concatenation products with molecular weights up to 20 kDa. Efficient ligation of peptoid foldamers may enable the chemical synthesis of biomimetic macromolecules capable of forming complex tertiary structures.