Merck
Localization of carbonic anhydrase IV in rat and human heart muscle.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (1998-06-20)
S Sender, B Decker, C D Fenske, W S Sly, N D Carter, G Gros
ABSTRACT
We investigated carbonic anhydrase IV (CA IV) in rat and human heart with immunohistochemical methods by both light and electron microscopy. In cryosections that were incubated with anti-CA IV/FITC, the capillaries showed a strong reaction for CA IV. In paraffin and semithin sections treated with anti-CA IV/ABC (avidin-biotin-peroxidase complex) blood vessels, capillaries, and sarcolemma (SL) were positively stained. By staining ultrathin sections with anti-CA IV/immunogold, CA IV could also be demonstrated at the latter two locations, including the specialized sarcolemmal structures intercalated discs, and T-tubules. In addition, by this method CA IV was seen to be associated with the sarcoplasmic reticulum (SR). The absence of immunostaining in SR and/or SL with some techniques probably indicates a problem of accessibility of the antigenic sites. In line with the immunohistochemical results, CA IV mRNA expression was visualized in both endothelial and muscle cells by in situ hybridization histochemistry.
Sigma-Aldrich
FluoroTag FITC Conjugation Kit