Merck
  • Synthesis and high-throughput screening of N-acetyl-beta-hexosaminidase inhibitor libraries targeting osteoarthritis.
Synthesis and high-throughput screening of N-acetyl-beta-hexosaminidase inhibitor libraries targeting osteoarthritis.
The Journal of organic chemistry (2004-09-11)
Junjie Liu, Mehdi M D Numa, Haitian Liu, Shi-Jung Huang, Pamela Sears, Alexander R Shikhman, Chi-Huey Wong
ABSTRACT
C1 Nitrogen iminocyclitols are potent inhibitors of N-acetyl-beta-hexosaminidases. Given hexosaminidases' important roles in osteoarthritis, we developed two straightforward and efficient syntheses of C1 nitrogen iminocyclitols from two readily available starting materials, D-mannosamine hydrochloride and the microbial oxidation product of fructose. A diversity-oriented synthetic strategy was then performed by coupling these core structures with various aldehydes, carboxylic acids, and alkynes to generate three separate libraries. High-throughput screening of the generated libraries with human N-acetyl-beta-hexosaminidases produced only moderate inhibitory activities. However, the synthetic approach and screening strategy for these compounds will be applied to develop new potent inhibitors of human N-acetyl-beta-hexosaminidases, particularly when combined with the structural information of these enzymes.
Sigma-Aldrich
D-Mannosamine hydrochloride, ≥98% (HPLC)
Pricing and availability is not currently available.