Merck
  • Carnitine effects on coenzyme A profiles in rat liver with hypoglycin inhibition of multiple dehydrogenases.
Carnitine effects on coenzyme A profiles in rat liver with hypoglycin inhibition of multiple dehydrogenases.
The American journal of physiology (1997-03-01)
Y K Lieu, B Y Hsu, W A Price, B E Corkey, C A Stanley
ABSTRACT
To examine the changes in coenzyme A profile and the possible corrective effects of carnitine supplementation in the genetic disorders of mitochondrial beta-oxidation, we carried out experiments using an inhibitor of multiple acyl-CoA dehydrogenase enzymes, methylenecyclopropaneacetic acid (MCPA), in rat hepatocytes. MCPA irreversibly inhibited ketone synthesis from straight-chain fatty acids (butyrate, octanoate, palmitate) and branched-chain fatty acids (alpha-ketoisocaproate) with a parallel 70-90% reduction of hepatocyte acetyl-CoA levels. Alone, MCPA or substrates halved free CoA levels to 15% of total CoA and doubled short- and medium-chain acyl-CoA levels to 30% of total CoA. With MCPA plus substrates combined, free CoA levels were 10% of total CoA, and short- and medium-chain acyl-CoA levels were 45% of total CoA. Comparable changes in CoA profiles were found in a patient with a severe genetic defect in beta-oxidation. Neither the suppression of ketogenesis nor the alterations in CoA profiles induced by MCPA inhibition could be corrected by carnitine supplementation.
Product Description
Supelco
(RS)-(Methylenecyclopropyl)acetic acid, analytical standard
Pricing and availability is not currently available.