Merck
  • Immune cells recruitment and activation by Tityus serrulatus scorpion venom.
Immune cells recruitment and activation by Tityus serrulatus scorpion venom.
Toxicon : official journal of the International Society on Toxinology (2011-09-07)
Eder M S Fialho, Márcia C G Maciel, Arlley C B Silva, Aramys S Reis, Anne Karine M Assunção, Thiare S Fortes, Lucilene A Silva, Rosane N M Guerra, Fabio H Kwasniewski, Flávia R F Nascimento
ABSTRACT
Despite several studies showed that the Tityus serrulatus scorpion venom (Tsv) induces an inflammatory response, just a few have investigated the effect of the venom on the immune response. Therefore, the aim of this study was to evaluate alterations of venom application on lymphoid organs and on the recruitment and activation of cells and also on the cytokine production. Swiss male mice (2-3 months, 20-25 g) received a non-lethal dose of crude Tsv (200 μg/kg), diluted in sterile PBS by subcutaneous route. Control animals received only sterile PBS. The animals were sacrificed after 30, 120 and 360 min. The inflammatory parameters studied were skin histology at the site of venom application, leukocyte count, and blood cytokine levels (IL-6, IL-10, and TNF-α). Inguinal lymph node, spleen and bone marrow cellularity was determined for evaluation of the Tsv effect on immune system organs. The results showed that Tsv caused no local inflammation, but it induced an increase of blood neutrophils and serum IL-6, TNF-α and IL-10. After 360 min of envenomation there was a reduction in the cells number from peritoneum and spleen, but there was an increase in the cell number from lymph nodes. In conclusion, the Tsv induces systemic alterations characterized by changes in the cell number in lymphoid organs, increase pro and anti-inflammatory cytokines.