Merck
  • Clickable Polyamine Derivatives as Chemical Probes for the Polyamine Transport System.
Clickable Polyamine Derivatives as Chemical Probes for the Polyamine Transport System.
Chembiochem : a European journal of chemical biology (2018-02-17)
Roeland Vanhoutte, Jan Pascal Kahler, Shaun Martin, Sarah van Veen, Steven H L Verhelst
ABSTRACT
Polyamines are essential for cell growth and differentiation, but their trafficking by the polyamine transport system is not fully understood. Herein, the synthesis of several azido-derivatized polyamines for easy conjugation by click chemistry is described. Attachment of a 4,4-difluoro-4-bora-3a,4a-diaza-s-indacene (BODIPY) dye gave fluorescent polyamine probes, which were tested in cell culture. The linear probe series showed superior cellular uptake compared with that of probes in which the dye was attached to a branch on one of the central amines. Interestingly, the linear probes accumulated rapidly in cancer cells (MCF-7), but not in nontumorigenic cells (MCF-10A). The fluorescent polyamine probes are therefore applicable to the study of polyamine trafficking, whereas the azido polyamines may be further utilized to transport cargo into cancer cells by exploiting the polyamine transport system.