Merck
  • KLF4 promotes milk fat synthesis by regulating the PI3K-AKT-mTOR pathway and targeting FASN activation in bovine mammary epithelial cells.
KLF4 promotes milk fat synthesis by regulating the PI3K-AKT-mTOR pathway and targeting FASN activation in bovine mammary epithelial cells.
iScience (2024-05-23)
Hong-Yu Wu, Zhong-Hao Ji, Wen-Yin Xie, Hai-Xiang Guo, Yi Zheng, Wei Gao, Bao Yuan
ABSTRACT
Milk fat is an important indicator for evaluating the quality of cow's milk. In this study, we used bovine mammary epithelial cells (BMECs) to investigate the role and molecular mechanism of KLF4 in the regulation of milk fat synthesis. The results showed that KLF4 was more highly expressed in mammary tissues of high-fat cows compared with low-fat cows. KLF4 positively regulated the expression of genes related to milk fat synthesis in BMECs, increasing intracellular triglycerides content, and KLF4 promoted milk fat synthesis by activating the PI3K-AKT-mTOR signaling pathway. Furthermore, the results of animal experiments also confirmed that knockdown of KLF4 inhibited milk fat synthesis. In addition, yeast one-hybrid assays and dual-luciferase reporter gene assays confirmed that KLF4 directly targets and binds to the fatty acid synthase (FASN) promoter region to promote FASN transcription. These results demonstrate that KLF4 is a key transcription factor for milk fat synthesis in BMECs.
Sigma-Aldrich
Hydrocortisone, Steroid hormone of the adrenal cortex with anti-inflammatory properties.