Merck
  • Pterostilbene-isothiocyanate reduces miR-21 level by impeding Dicer-mediated processing of pre-miR-21 in 5-fluorouracil and tamoxifen-resistant human breast cancer cell lines.
Pterostilbene-isothiocyanate reduces miR-21 level by impeding Dicer-mediated processing of pre-miR-21 in 5-fluorouracil and tamoxifen-resistant human breast cancer cell lines.
3 Biotech (2023-05-19)
Viney Kumar, Swati Haldar, Saakshi Saini, Souvik Ghosh, Poonam Dhankhar, Partha Roy
ABSTRACT
Converging evidences identifies that microRNA-21 (miR-21) is responsible for drug resistance in breast cancer. This study aims to evaluate the miR-21-modulatory potential of a hybrid compound, pterostilbene-isothiocyanate (PTER-ITC), in tamoxifen-resistant MCF-7 (TR/MCF-7) and 5-fluorouracil-resistant MDA-MB 231 (5-FUR/MDA-MB 231) breast cancer cell lines, established by repeated exposure to gradually increasing the concentrations of tamoxifen and 5-fluorouracil, respectively. The outcome of this study shows that PTER-ITC effectively reduced the TR/MCF-7 (IC50: 37.21 µM) and 5-FUR/MDA-MB 231 (IC50: 47.00 µM) cell survival by inducing apoptosis, inhibiting cell migration, colony and spheroid formations in TR/MCF-7 cells, and invasiveness of 5-FUR/MDA-MB 231 cells. Most importantly, PTER-ITC significantly reduced the miR-21 expressions in these resistant cell lines. Moreover, the downstream tumor suppressor target gene of miR-21 such as PTEN, PDCD4, TIMP3, TPM1, and Fas L were upregulated after PTER-ITC treatment, as observed from transcriptional (RT-qPCR) and translational (immunoblotting) data. In silico and miR-immunoprecipitation (miR-IP) results showed reduced Dicer binding to pre-miR-21, after PTER-ITC treatment, indicating inhibition of miR-21 biogenesis. Collectively, the significance of this study is indicated by preliminary evidence for miR-21-modulatory effects of PTER-ITC that highlights the potential of this hybrid compound as an miR-21-targeting therapeutic agent.
Sigma-Aldrich
Tamoxifen Citrate, Tamoxifen is a potent synthetic anti-estrogenic agent.
Pricing and availability is not currently available.