Merck
  • RNA polymerase II pausing temporally coordinates cell cycle progression and erythroid differentiation.
RNA polymerase II pausing temporally coordinates cell cycle progression and erythroid differentiation.
Developmental cell (2023-08-17)
Danya J Martell, Hope E Merens, Alexis Caulier, Claudia Fiorini, Jacob C Ulirsch, Robert Ietswaart, Karine Choquet, Giovanna Graziadei, Valentina Brancaleoni, Maria Domenica Cappellini, Caroline Scott, Nigel Roberts, Melanie Proven, Noémi B A Roy, Christian Babbs, Douglas R Higgs, Vijay G Sankaran, L Stirling Churchman
ABSTRACT
Controlled release of promoter-proximal paused RNA polymerase II (RNA Pol II) is crucial for gene regulation. However, studying RNA Pol II pausing is challenging, as pause-release factors are almost all essential. In this study, we identified heterozygous loss-of-function mutations in SUPT5H, which encodes SPT5, in individuals with β-thalassemia. During erythropoiesis in healthy human cells, cell cycle genes were highly paused as cells transition from progenitors to precursors. When the pathogenic mutations were recapitulated by SUPT5H editing, RNA Pol II pause release was globally disrupted, and as cells began transitioning from progenitors to precursors, differentiation was delayed, accompanied by a transient lag in erythroid-specific gene expression and cell cycle kinetics. Despite this delay, cells terminally differentiate, and cell cycle phase distributions normalize. Therefore, hindering pause release perturbs proliferation and differentiation dynamics at a key transition during erythropoiesis, identifying a role for RNA Pol II pausing in temporally coordinating the cell cycle and erythroid differentiation.