Merck
  • A Novel Inactive Isoform with a Restored Reading Frame Is Expressed from the Human Interferon Lambda 4 TT Allele at rs368234815.
A Novel Inactive Isoform with a Restored Reading Frame Is Expressed from the Human Interferon Lambda 4 TT Allele at rs368234815.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research (2023-03-08)
Seema Bharatiya, Aditya Agarwal, Sreedhar Chinnaswamy
ABSTRACT
The TT allele of the dinucleotide variant rs368234815 (TT/ΔG) abolishes the open reading frame (ORF) created by the ancestral ΔG allele of the human interferon lambda 4 (IFNL4) gene, thus preventing the expression of a functional IFN-λ4 protein. While probing the expression of IFN-λ4 in human peripheral blood mononuclear cells (PBMCs), using a monoclonal antibody that binds to the C-terminus of IFN-λ4, surprisingly, we observed that PBMCs obtained from TT/TT genotype individuals could also express proteins that reacted with the IFN-λ4-specific antibody. We confirmed that these products did not emanate from the IFNL4 paralog, IF1IC2 gene. Using cell lines and overexpressing human IFNL4 gene constructs, we obtained evidence from Western blots to show that the TT allele could express a protein that reacted with the IFN-λ4 C-terminal-specific antibody. It had a molecular weight similar if not identical to IFN-λ4 expressed from the ΔG allele. Furthermore, the same start and stop codons used by the ΔG allele were used to express the novel isoform from the TT allele suggesting that a restoration of the ORF had occurred in the body of the mRNA. However, this TT allele isoform did not induce any IFN-stimulated gene expression. Our data do not support a ribosomal frameshift that leads to the expression of this new isoform, implying that an alternate splicing event may be responsible. An N-terminal-specific monoclonal antibody did not react with the novel protein isoform suggesting that the alternate splicing event likely occurs beyond exon 2. The new isoform is glycosylated similar to the functional IFN-λ4 and is also secreted. Furthermore, we show that the ΔG allele can also potentially express a similarly frameshifted isoform. The splicing event that leads to the generation of these novel isoforms and their functional significance remains to be elucidated.
Sigma-Aldrich
Anti-Interferon Lambda 4, clone 4G1 Antibody, clone 4G1, 1 mg/mL, from mouse
Pricing and availability is not currently available.