Merck
  • Modulation of Breast Cancer Cell FASN Expression by Obesity-Related Systemic Factors.
Modulation of Breast Cancer Cell FASN Expression by Obesity-Related Systemic Factors.
Breast cancer : basic and clinical research (2022-08-30)
Bryan McClellan, Tommy Pham, Brittany Harlow, Gabby Lee, Duan Quach, Christopher Jolly, Andrew Brenner, Linda deGraffenried
ABSTRACT
The objective of this study is to determine the impact of exposure to obesity-related systemic factors on fatty acid synthase enzyme (FASN) expression in breast cancer cells. MCF-7 breast cancer cells were exposed to sera from patients having obesity or not having obesity and subjected to quantitative reverse transcription polymerase chain reaction (RT-qPCR). Subsequent MTT and colony-forming assays using both MCF-7 and T-47D cells exposed to sera and treated with or without FASN inhibitor, TVB-3166, were used. MCF-7 cells were then treated with insulin and the sterol regulatory element-binding protein (SREBP) processing inhibitor, betulin, prior to analysis of FASN expression by quantitative RT-qPCR and western blot. Insulin-induced SREBP-FASN promoter binding was analyzed by chromatin immunoprecipitation with an anti-SREBP antibody. In response to sera exposure (body mass index [BMI] >30) there was an increase in FASN expression in breast cancer cells. Furthermore, treatment with the FASN inhibitor, TVB-3166, resulted in a decreased breast cancer cell survival and proliferation while increasing apoptosis upon sera exposure (BMI >30). Insulin-exposed MCF-7 cells exhibited an increased FASN messenger RNA and protein expression, which is abrogated upon SREBP inhibition. In addition, insulin exposure induced enhanced SREBP binding to the FASN promoter. Our results implicate FASN as a potential mediator of obesity-induced breast cancer aggression and a therapeutic target of patients with obesity-induced breast cancer.
Sigma-Aldrich
SREBP Processing Inhibitor, Betulin, The SREBP Processing Inhibitor, Betulin, also referenced under CAS 473-98-3, controls the biological activity of SCAP (SREBP cleavage activating protein). This small molecule/inhibitor is primarily used for Cell Signaling applications.
Pricing and availability is not currently available.