Merck
  • Reconfiguration of genomic anchors upon transcriptional activation of the human major histocompatibility complex.
Reconfiguration of genomic anchors upon transcriptional activation of the human major histocompatibility complex.
Genome research (2008-10-14)
Diego Ottaviani, Elliott Lever, Richard Mitter, Tania Jones, Tim Forshew, Rossitza Christova, Eleni M Tomazou, Vardhman K Rakyan, Stephen A Krawetz, Adrian E Platts, Badmavady Segarane, Stephan Beck, Denise Sheer
ABSTRACT
The folding of chromatin into topologically constrained loop domains is essential for genomic function. We have identified genomic anchors that define the organization of chromatin loop domains across the human major histocompatibility complex (MHC). This locus contains critical genes for immunity and is associated with more diseases than any other region of the genome. Classical MHC genes are expressed in a cell type-specific pattern and can be induced by cytokines such as interferon-gamma (IFNG). Transcriptional activation of the MHC was associated with a reconfiguration of chromatin architecture resulting from the formation of additional genomic anchors. These findings suggest that the dynamic arrangement of genomic anchors and loops plays a role in transcriptional regulation.
Sigma-Aldrich
JumpStart Taq ReadyMix, Complete optimized reagent for hot-start PCR at 2X concentration
Pricing and availability is not currently available.