Merck
  • The NFATc1/P2X7 receptor relationship in human intervertebral disc cells.
The NFATc1/P2X7 receptor relationship in human intervertebral disc cells.
Frontiers in cell and developmental biology (2024-04-19)
Maria Pina Notarangelo, Letizia Penolazzi, Elisabetta Lambertini, Simonetta Falzoni, Pasquale De Bonis, Cristina Capanni, Francesco Di Virgilio, Roberta Piva
ABSTRACT
A comprehensive understanding of the molecules that play key roles in the physiological and pathological homeostasis of the human intervertebral disc (IVD) remains challenging, as does the development of new therapeutic treatments. We recently found a positive correlation between IVD degeneration (IDD) and P2X7 receptor (P2X7R) expression increases both in the cytoplasm and in the nucleus. Using immunocytochemistry, reverse transcription PCR (RT-PCR), overexpression, and chromatin immunoprecipitation, we found that NFATc1 and hypoxia-inducible factor-1α (HIF-1α) are critical regulators of P2X7R. Both transcription factors are recruited at the promoter of the P2RX7 gene and involved in its positive and negative regulation, respectively. Furthermore, using the proximity ligation assay, we revealed that P2X7R and NFATc1 form a molecular complex and that P2X7R is closely associated with lamin A/C, a major component of the nuclear lamina. Collectively, our study identifies, for the first time, P2X7R and NFATc1 as markers of IVD degeneration and demonstrates that both NFATc1 and lamin A/C are interaction partners of P2X7R.
Sigma-Aldrich
Anti-P2X7 Receptor Antibody, Chemicon®, from rabbit