Merck
  • Formyl peptide receptors: a promiscuous subfamily of G protein-coupled receptors controlling immune responses.
Formyl peptide receptors: a promiscuous subfamily of G protein-coupled receptors controlling immune responses.
Cytokine & growth factor reviews (2006-11-07)
Isabelle Migeotte, David Communi, Marc Parmentier
ABSTRACT
The formyl peptide receptor (FPR) family is involved in host defence against pathogens, but also in sensing internal molecules that may constitute signals of cellular dysfunction. It includes three subtypes in human and other primates. FPR responds to formyl peptides derived from bacterial and mitochondrial proteins. FPRL1 displays a large array of exogenous and endogenous ligands, including the chemokine variant sCKbeta8-1, the neuroprotective peptide humanin, and lipoxin A4. Two high affinity agonists (F2L and humanin) were recently described for FPRL2. In mouse, eight FPR-related receptors have been described. Fpr1 is the ortholog of human FPR, while fpr2 appears to share many ligands with human FPRL1. Altogether, the physiological role of the FPR family is still incompletely understood, due in part to the large variety of ligands, the redundancy with other chemoattractant agents, and the lack of clear orthologs between human and mouse receptors. Newly developed tools will allow to study further this family of receptors.
Sigma-Aldrich
Cathepsin G, Human Neutrophil, Cathepsin G, Human Neutrophil, CAS 107200-92-0, is a purified native cathepsin G. Acts as a potent agonist of human platelet activation leading to their aggregation.
Pricing and availability is not currently available.