Merck
  • Hypothalamic neuronal outputs transmit sensorimotor signals at the onset of locomotor initiation.
Hypothalamic neuronal outputs transmit sensorimotor signals at the onset of locomotor initiation.
iScience (2023-11-29)
Ekaterina Martianova, Renata Sadretdinova, Alicia Pageau, Nikola Pausic, Tommy Doucet Gentiletti, Danahé Leblanc, Arturo Marroquin Rivera, Benoît Labonté, Christophe D Proulx
ABSTRACT
The lateral hypothalamus (LH) plays a critical role in sensory integration to organize behavior responses. However, how projection-defined LH neuronal outputs dynamically transmit sensorimotor signals to major downstream targets to organize behavior is unknown. Here, using multi-fiber photometry, we show that three major LH neuronal outputs projecting to the dorsal raphe nucleus (DRN), ventral tegmental area (VTA), and lateral habenula (LHb) exhibit significant coherent activity in mice engaging sensory-evoked or self-initiated motor responses. Increased activity at LH axon terminals precedes movement initiation during active coping responses and the activity of serotonin neurons and dopamine neurons. The optogenetic activation of LH axon terminals in either of the DRN, VTA, or LHb was sufficient to increase motor initiation but had different effects on passive avoidance and sucrose consumption. Our findings support the complementary role of three projection-defined LH neuronal outputs in the transmission of sensorimotor signals to major downstream regions at movement onset.
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from sheep
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Tryptophan Hydroxylase Antibody, Chemicon®, from sheep
Pricing and availability is not currently available.