Merck
  • Growth hormone (GH) and growth hormone release factor (GRF) modulate the immune response in the SHK-1 cell line and leukocyte cultures of head kidney in Atlantic salmon.
Growth hormone (GH) and growth hormone release factor (GRF) modulate the immune response in the SHK-1 cell line and leukocyte cultures of head kidney in Atlantic salmon.
General and comparative endocrinology (2020-10-04)
J P Pontigo, L Vargas-Chacoff
ABSTRACT
To clarify the role of growth hormone (GH) in the immune system of fish, we examine the comparative effect of GH and Growth Hormone Release Factor (GRF) on leukocytes culture of the head kidney of Atlantic salmon and the SHK-1 cell line. There are studies that associate the growth hormone (GH) / insulin-like growth factor (IGF) axis with the immune regulation of fish. However, there is no evidence that GH and GRF stimulate Atlantic salmon leukocyte cell lines, where there areńt reports on expression changes in different immune response markers. Thus, we investigated the effect of GH and GRF in Atlantic salmon leukocytes extracted from head kidney and the SHK-1 cell line on the different immune response markers such as: NLRC5, NLRC3, IL-1β, TNF-α, and IL-8 through qPCR. Our data suggest that GH increases the expression of NLRC5, NLRC3, and IL-1β mainly at 16 h post-stimulation in Atlantic salmon leukocytes. This indicates differential regulation between the two models used, helping us to better understand the independent action of GH on the immune system and the GH / IGF axis for future research.