Merck
  • METTL3-Mediated m6A Modification Controls Splicing Factor Abundance and Contributes to Aggressive CLL.
METTL3-Mediated m6A Modification Controls Splicing Factor Abundance and Contributes to Aggressive CLL.
Blood cancer discovery (2023-04-18)
Yiming Wu, Meiling Jin, Mike Fernandez, Kevyn L Hart, Aijun Liao, Xinzhou Ge, Stacey M Fernandes, Tinisha McDonald, Zhenhua Chen, Daniel Röth, Lucy Y Ghoda, Guido Marcucci, Markus Kalkum, Raju K Pillai, Alexey V Danilov, Jingyi Jessica Li, Jianjun Chen, Jennifer R Brown, Steven T Rosen, Tanya Siddiqi, Lili Wang
ABSTRACT
RNA splicing dysregulation underlies the onset and progression of cancers. In chronic lymphocytic leukemia (CLL), spliceosome mutations leading to aberrant splicing occur in ∼20% of patients. However, the mechanism for splicing defects in spliceosome-unmutated CLL cases remains elusive. Through an integrative transcriptomic and proteomic analysis, we discover that proteins involved in RNA splicing are posttranscriptionally upregulated in CLL cells, resulting in splicing dysregulation. The abundance of splicing complexes is an independent risk factor for poor prognosis. Moreover, increased splicing factor expression is highly correlated with the abundance of METTL3, an RNA methyltransferase that deposits N6-methyladenosine (m6A) on mRNA. METTL3 is essential for cell growth in vitro and in vivo and controls splicing factor protein expression in a methyltransferase-dependent manner through m6A modification-mediated ribosome recycling and decoding. Our results uncover METTL3-mediated m6A modification as a novel regulatory axis in driving splicing dysregulation and contributing to aggressive CLL. METTL3 controls widespread splicing factor abundance via translational control of m6A-modified mRNA, contributes to RNA splicing dysregulation and disease progression in CLL, and serves as a potential therapeutic target in aggressive CLL. See related commentary by Janin and Esteller, p. 176. This article is highlighted in the In This Issue feature, p. 171.
Sigma-Aldrich
Anti-METTL14 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-METTL16 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution