Merck
  • IGF2BP2 promotes ovarian cancer growth and metastasis by upregulating CKAP2L protein expression in an m6 A-dependent manner.
IGF2BP2 promotes ovarian cancer growth and metastasis by upregulating CKAP2L protein expression in an m6 A-dependent manner.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2023-09-04)
Yaqian Shi, Xueyou Xiong, Yu Sun, Zhe Geng, Xiyi Chen, Xin Cui, Juan Lv, Lili Ge, Xuemei Jia, Juan Xu
ABSTRACT
Ovarian cancer (OC) is the second leading cause of gynecological cancer-related death in women worldwide. N6-methyladenosine (m6 A) is the most abundant internal modification in eukaryotic RNA. Human insulin-like growth factor 2 mRNA-binding protein 2 (IGF2BP2), an m6 A reader, can enhance mRNA stability and promote translation by recognizing m6 A modifications. Its tumor-promoting effects have been demonstrated in several cancers. However, the roles of m6 A modification and IGF2BP2 in OC remain unclear. Here, by using methylated RNA immunoprecipitation sequencing, we demonstrated that there is widespread dysregulation of m6 A modification in OC tissues. The m6 A modification and the mRNA and protein levels of IGF2BP2 were significantly elevated in OC. Overexpression of IGF2BP2 facilitated OC cell proliferation, migration, and invasion in vitro and accelerated tumor growth and metastasis in vivo. While IGF2BP2-knockdown showed the opposite effect. Mechanistically, we identified cytoskeleton-associated protein 2-like (CKAP2L) as a target of IGF2BP2. IGF2BP2 promoted CKAP2L translation dependent on m6 A modification, rather than affecting mRNA and protein stability. Overexpression of CKAP2L rescued the tumor-suppressive effect of IGF2BP2 knockdown in OC cells. In conclusion, this study revealed the potential role of IGF2BP2 in tumor progression, at least partially via promoting the translation of CKAP2L in an m6 A-dependent manner.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-N6-methyladenosine (m6A) Antibody, from rabbit
Sigma-Aldrich
IgG, Rabbit, Rabbit IgG Polyclonal Antibody validated for use in ELISA.