Merck
  • Behavioral and Transcriptional Effects of Short or Prolonged Fasting on the Memory Performances of Lymnaea stagnalis.
Behavioral and Transcriptional Effects of Short or Prolonged Fasting on the Memory Performances of Lymnaea stagnalis.
Neuroendocrinology (2022-10-13)
Veronica Rivi, Cristina Benatti, Pietro Actis, Fabio Tascedda, Johanna Maria Catharina Blom
ABSTRACT
The Garcia effect, a solid learning paradigm, was used to investigate the molecular and behavioral effects induced by different lengths of fasting on the cognitive functions in the pond snail Lymnaea stagnalis, a valid model system. Three experimental groups were used: moderately hungry snails, food-deprived for 1 day (D1 snails), severely hungry snails (D5 snails), fasting for 5 days, and satiated snails with ad libitum access to food (AL snails). In the Garcia effect, a single pairing of an appetitive stimulus with a heat stressor results in a learned taste-specific negative hedonic shift. D5 snails were injected with bovine insulin and D1 snails with the insulin receptor antibody (Ab). As a control group, AL snails were injected with saline. Gene expression analyses were performed by real-time PCR in snails' central nervous system (CNS). AL snails are "average learners," D1 snails are the best performers, whereas the D5 ones do not show the Garcia effect. Severely fasting snails injected with insulin 3 h before the training procedure show the Garcia effect, whereas injecting 1-day fasting snails with insulin receptor Ab blocks their ability to express memory. The differences in memory performances are associated with changes in the expression levels of selected targets involved in neuronal plasticity, energy homeostasis, and stress response. Our results suggest that short-term fasting creates an optimal internal state in L. stagnalis' CNS, allowing a spike in insulin release and an upregulation of genes involved in neuroplasticity. Long-term fasting, instead, upregulates genes involved in energy homeostasis and animal survival.