Merck
  • Targeting of SLC25A22 boosts the immunotherapeutic response in KRAS-mutant colorectal cancer.
Targeting of SLC25A22 boosts the immunotherapeutic response in KRAS-mutant colorectal cancer.
Nature communications (2023-08-05)
Qiming Zhou, Yao Peng, Fenfen Ji, Huarong Chen, Wei Kang, Lam-Shing Chan, Hongyan Gou, Yufeng Lin, Pingmei Huang, Danyu Chen, Qinyao Wei, Hao Su, Cong Liang, Xiang Zhang, Jun Yu, Chi Chun Wong
ABSTRACT
KRAS is an important tumor intrinsic factor driving immune suppression in colorectal cancer (CRC). In this study, we demonstrate that SLC25A22 underlies mutant KRAS-induced immune suppression in CRC. In immunocompetent male mice and humanized male mice models, SLC25A22 knockout inhibits KRAS-mutant CRC tumor growth with reduced myeloid derived suppressor cells (MDSC) but increased CD8+ T-cells, implying the reversion of mutant KRAS-driven immunosuppression. Mechanistically, we find that SLC25A22 plays a central role in promoting asparagine, which binds and activates SRC phosphorylation. Asparagine-mediated SRC promotes ERK/ETS2 signaling, which drives CXCL1 transcription. Secreted CXCL1 functions as a chemoattractant for MDSC via CXCR2, leading to an immunosuppressive microenvironment. Targeting SLC25A22 or asparagine impairs KRAS-induced MDSC infiltration in CRC. Finally, we demonstrate that the targeting of SLC25A22 in combination with anti-PD1 therapy synergizes to inhibit MDSC and activate CD8+ T cells to suppress KRAS-mutant CRC growth in vivo. We thus identify a metabolic pathway that drives immunosuppression in KRAS-mutant CRC.
Sigma-Aldrich
Anti-SLC25A22 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.