Merck
  • Central Irisin Signaling Is Required for Normal Timing of Puberty in Female Mice.
Central Irisin Signaling Is Required for Normal Timing of Puberty in Female Mice.
Endocrinology (2022-12-13)
Caroline Decourt, Maggie C Evans, Megan A Inglis, Greg M Anderson
ABSTRACT
Timing of puberty requires exquisite coordination of genes, hormones, and brain circuitry. An increasing level of body adiposity, signaled to the brain via the fat-derived hormone leptin, is recognized as a major factor controlling puberty onset. However, it is clear that leptin is not the only metabolic cue regulating puberty, and that developmental regulation of this process also involves tissues other than adipose, with muscle development potentially playing a role in the timing of puberty. The proteolytic processing of fibronectin type 3 domain-containing protein 5 (FNDC5) releases a hormone, irisin. Irisin is primarily produced by muscle and is released into circulation, where levels increase dramatically as puberty approaches. We investigated the effects of a global deletion of the Fndc5 gene on pubertal timing. The absence of irisin induced a delay in puberty onset in female knockout mice compared with controls, without affecting body weight or gonadotropin-releasing hormone (GnRH) neuronal density. We next treated pre-pubertal wild-type male and female mice with an irisin receptor antagonist, cilengitide, for 7 days and observed a delay in first estrus occurrence compared to vehicle-treated control mice. Male puberty timing was unaffected. Next, we deleted the irisin receptor (integrin subunit alpha V) in all forebrain neurons and found a delay in the occurrence of first estrus in knockout females compared to controls. Taken together, these data suggest irisin plays a role in the timing of puberty onset in female mice via a centrally mediated mechanism.
Sigma-Aldrich
Cilengitide trifluoroacetic acid salt, ≥95% (HPLC)
Pricing and availability is not currently available.