Merck
  • MAIT cell inhibition promotes liver fibrosis regression via macrophage phenotype reprogramming.
MAIT cell inhibition promotes liver fibrosis regression via macrophage phenotype reprogramming.
Nature communications (2023-04-04)
Morgane Mabire, Pushpa Hegde, Adel Hammoutene, Jinghong Wan, Charles Caër, Rola Al Sayegh, Mathilde Cadoux, Manon Allaire, Emmanuel Weiss, Tristan Thibault-Sogorb, Olivier Lantz, Michèle Goodhardt, Valérie Paradis, Pierre de la Grange, Hélène Gilgenkrantz, Sophie Lotersztajn
ABSTRACT
Recent data have shown that liver fibrosis can regress even at later stages of cirrhosis and shifting the immune response from pro-inflammatory towards a resolutive profile is considered as a promising option. The immune regulatory networks that govern the shift of the inflammatory phenotype and thus potential reversal of liver fibrosis are lesser known. Here we show that in precision-cut human liver slices obtained from patients with end-stage fibrosis and in mouse models, inhibiting Mucosal-Associated Invariant T (MAIT) cells using pharmacological or antibody-driven approaches, limits fibrosis progression and even regresses fibrosis, following chronic toxic- or non-alcoholic steatohepatitis (NASH)-induced liver injury. Mechanistic studies, combining RNA sequencing, in vivo functional studies (performed in male mice) and co-culture experiments indicate that disruption of the MAIT cell-monocyte/macrophage interaction results in resolution of fibrosis both by increasing the frequency of restorative Ly6Clo at the expenses of pro-fibrogenic Ly6Chi monocyte-derived macrophages and promoting an autophagic phenotype in both subsets. Thus, our data show that MAIT cell activation and the consequential phenotype shift of liver macrophages are important pathogenic features of liver fibrosis and could be targeted by anti-fibrogenic therapy.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Acetyl-6-formylpterin, ≥95% (HPLC)